Invitrogen
Manufacturer Code:MA524080
Catalog # PIMA524080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 344015 |
Immunogen | E. coli-derived recombit mouse S100A10 Met1-Lys97 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 42C ANX2L ANX2LG CAL1L CLP11 Ca&lsqb1] GP11 P11 p10 S100 calcium binding protein A10 (annexin II ligand calpactin I light polypeptide (p11)) S100 calcium-binding protein A10 (annexin II ligand calpactin I light polypeptide (p11)) annexin II ligand calpactin I light polypeptide calpactin I light chain; calcium binding protein A11 (calgizzarin); Calpactin I light chain; Calpactin-1 light chain; Cellular ligand of annexin II; p10 protein; p11; Protein S100-A10; S100 calcium binding protein A10 (calgizzarin); S100 calcium-binding protein A10
Gene Aliases: 42C; AA409961; AL024248; CAL12; Cal1l; CLP11; p10; p11; S100a10
UniProt ID: (Mouse) P08207
Entrez Gene ID: (Mouse) 20194
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.