R&D Systems
Manufacturer Code:FAB2377R100UG
Catalog # FAB2377R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 344015 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse S100A10 Met1-Lys97 Accession # P08207 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: annexin II ligand calpactin I light polypeptide ANX2L ANX2LG CAL1LGP1142C Calpactin I light chain Calpactin-1 light chain Cellular ligand of annexin II CLP11Ca[1] MGC111133 p10 p10 protein p11 protein S100-A10 S100 calcium binding protein A10 S100 calcium binding protein A10 (annexin II ligand calpactin I lightpolypeptide (p11)) S100 calcium-binding protein A10 S100 calcium-binding protein A10 (annexin II ligand calpactin I lightpolypeptide (p11)); annexin II ligand, calpactin I, light polypeptide; annexin II tetramer (AIIt) p11 subunit; Calpactin I light chain; Calpactin-1 light chain; Cellular ligand of annexin II; p10 protein; p11; Protein S100-A10; S100 calcium binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S100 calcium-binding protein A10; S100 calcium-binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11))
Gene Aliases: 42C; ANX2L; ANX2LG; Ca[1]; CAL1L; CLP11; GP11; p10; P11; S100A10
UniProt ID: (Human) P60903
Entrez Gene ID: (Human) 6281
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.