Proteintech
Manufacturer Code:666161IG150UL
Catalog # 50-173-7034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G10F4 |
Immunogen | S100 beta Fusion Protein Ag7440 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NEF Protein S100 B S 100 protein beta chain S 100 protein subunit beta S100 S100 calcium binding protein B S100B S100beta; Protein S100-B; S-100 calcium-binding protein, beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S100 protein, beta polypeptide, neural
Gene Aliases: NEF; S100; S100-B; S100B; S100beta; S100P
UniProt ID: (Human) P04271, (Rat) P04631
Entrez Gene ID: (Human) 6285, (Pig) 100623510, (Rat) 25742
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.