Invitrogen
Manufacturer Code:MA120103
Catalog # PIMA120103
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6G1 |
Immunogen | Human S-100 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bpb NEF S 100; Protein S100-A1; Protein S100-B; S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 alpha; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein, beta (neural); S100 protein, alpha polypeptide
Gene Aliases: NEF; S100; S100-alpha; S100-B; S100A; S100A1; S100B; S100beta
UniProt ID: (Human) P23297, (Human) P04271
Entrez Gene ID: (Human) 6271, (Human) 6285
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.