Invitrogen
Manufacturer Code:MA512966
Catalog # PIMA512966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C4.9 |
Immunogen | Purified bovine brain S100 protein |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Protein S100-A1; Protein S100-B; S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein alpha chain S-100 protein subunit alpha S100 alpha S100 calcium-binding protein A1 S100 protein alpha polypeptide protein S100-A1; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 alpha; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural
Gene Aliases: AI266795; AI850290; Bpb; NEF; S100; S100-alpha; S100-B; S100A; S100A1; S100B; S100beta; S100P
UniProt ID: (Bovine) P02639, (Human) P23297, (Human) P04271, (Bovine) P02638, (Rat) P35467, (Mouse) P56565, (Mouse) P50114, (Rat) P04631
Entrez Gene ID: (Bovine) 528735, (Human) 6271, (Human) 6285, (Bovine) 525716, (Rat) 295214, (Mouse) 20193, (Mouse) 20203, (Rat) 25742
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.