Invitrogen
Manufacturer Code:PA5115860
Catalog # PIPA5115860
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human S100B corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI266795 AI850290 Bpb hypothetical protein LOC525716 NEF Protein S100-A1 protein S100-B S 100 S100 S100 alpha S100 calcium binding protein A1 S100 calcium binding protein B S100 calcium-binding protein A1 S100 calcium-binding protein B S100 calcium-binding protein beta (neural) S100 calcium-binding protein beta (neural) S-100 calcium-binding protein beta chain S-100 protein alpha chain S-100 protein beta chain S-100 protein subunit alpha S-100 protein subunit beta S100 protein alpha polypeptide S100 protein beta polypeptide S100 protein beta polypeptide neural S100A S100A1 S100alpha S100-alpha S100B S100-B S100beta S100P; Protein S100-A1; Protein S100-B; S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 alpha; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S100 protein, alpha polypeptide; S100 protein, beta polypeptide, neural
Gene Aliases: AI266795; AI850290; Bpb; NEF; S100; S100-alpha; S100-B; S100A; S100A1; S100B; S100beta; S100P
UniProt ID: (Human) P23297, (Human) P04271, (Rat) P35467, (Mouse) P56565, (Mouse) P50114, (Rat) P04631
Entrez Gene ID: (Human) 6271, (Human) 6285, (Rat) 295214, (Mouse) 20193, (Mouse) 20203, (Rat) 25742
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.