Novus Biologicals
Manufacturer Code:NB1002385SS
Catalog # NB1002385SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PDS-1 |
Immunogen | Porcine retinal S-arrestin [Swiss-Prot# P79260] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 48 kDa protein; arrestin 1; arrestin 1 RP4748 kDa protein S-antigen retina and pineal gland (arrestin) S-arrestin visual arrestin; Retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; Rod photoreceptor arrestin; S-AG; S-antigen; retina and pineal gland (arrestin); S-arrestin
Gene Aliases: RP47; S-AG; SAG
UniProt ID: (Human) P10523
Entrez Gene ID: (Human) 6295
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.