Invitrogen
Manufacturer Code:MA15872
Catalog # 01-674-950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PDS-1 |
Immunogen | Porcine S-antigen from Balb/c Ag 8.653 cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 48 kDa protein; Arrestin; arrestin 1; arrestin1; photoreceptor protein/alleged cGMP phosphodiesterase activation modulator; retinal S antigen; Retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; Rod photoreceptor arrestin; S-AG; S-antigen; S-antigen; retina and pineal gland (arrestin); S-arrestin; S-arrestin short form
Gene Aliases: RP47; S-AG; SAG; SAGMR; SANTI
UniProt ID: (Pig) P79260, (Bovine) P08168, (Human) P10523, (Rat) P15887
Entrez Gene ID: (Pig) 397151, (Bovine) 280922, (Human) 6295, (Rat) 25539
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.