Invitrogen
Manufacturer Code:PA579953
Catalog # PIPA579953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human Retinal S antigen recombit protein (Position: R193-E405). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 48 kDa protein; arrestin 1; Retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; Rod photoreceptor arrestin; S-AG; S-antigen; S-antigen; retina and pineal gland (arrestin); S-arrestin; S-arrestin 48 kDa protein Retinal S-antigen S-AG Rod photoreceptor arrestin SAG; visual arrestin 1
Gene Aliases: A930001K18Rik; Arr1; arrestin; Irbp; RP47; S-AG; SAG; SAGMR; SANTI
UniProt ID: (Human) P10523, (Rat) P15887, (Mouse) P20443
Entrez Gene ID: (Human) 6295, (Rat) 25539, (Mouse) 20215
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.