Novus Biologicals
Manufacturer Code:NBP299540B
Catalog # NB061488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse Ryk. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D3S3195 EC 2.7.10 EC 2.7.10.1 hydroxyaryl-protein kinase JTK5 JTK5A JTK5A protein tyrosine kinase RYK receptor-like tyrosine kinase RYK1 tyrosine-protein kinase RYK; hydroxyaryl-protein kinase; JTK5A protein tyrosine kinase; Tyrosine-protein kinase RYK
Gene Aliases: D3S3195; JTK5; JTK5A; RYK; RYK1
UniProt ID: (Human) P34925
Entrez Gene ID: (Human) 6259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.