R&D Systems
Manufacturer Code:AF4649SP
Catalog # AF4649SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Ryk Arg35-Arg211 Accession # Q01887 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: D3S3195 EC 2.7.10 EC 2.7.10.1 hydroxyaryl-protein kinase JTK5 JTK5A JTK5A protein tyrosine kinase RYK receptor-like tyrosine kinase RYK1 tyrosine-protein kinase RYK; hydroxyaryl-protein kinase; JTK5A protein tyrosine kinase; Tyrosine-protein kinase RYK
Gene Aliases: D3S3195; JTK5; JTK5A; RYK; RYK1
UniProt ID: (Human) P34925
Entrez Gene ID: (Human) 6259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.