Novus Biologicals
Manufacturer Code:NBP280143DL594
Catalog # NB013282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | C3-33 |
Immunogen | Canine cardiac ryanodine receptor (RyR2) |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: arrhythmogenic right ventricular dysplasia 2 ARVC2 ARVD2 cardiac muscle ryanodine receptor-calcium release channel Cardiac muscle-type ryanodine receptor cardiac-type ryanodine receptor hRYR-2 islet-type ryanodine receptor kidney-type ryanodine receptor ryanodine receptor 2 ryanodine receptor 2 (cardiac) RyR2 RYR-2 type 2 ryanodine receptor VTSIP; Cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac-type ryanodine receptor; islet-type ryanodine receptor; kidney-type ryanodine receptor; Ryanodine receptor 2; ryanodine receptor 2 (cardiac); RYR-2; Type 2 ryanodine receptor
Gene Aliases: ARVC2; ARVD2; RyR; RYR-2; RYR2; VTSIP
UniProt ID: (Human) Q92736
Entrez Gene ID: (Human) 6262
Molecular Function:
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.