Invitrogen
Manufacturer Code:PA587416
Catalog # PIPA587416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a sequence within amino acids 4850-4950 of human RYR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium release channel; Cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac-type ryanodine receptor; islet-type ryanodine receptor; kidney-type ryanodine receptor; Ryanodine receptor 2; ryanodine receptor 2 (cardiac); ryanodine receptor type 2; ryanodine receptor type II; RYR-2; Type 2 ryanodine receptor
Gene Aliases: 9330127I20Rik; ARVC2; ARVD2; RyR; RYR-2; RYR2; VTSIP
UniProt ID: (Human) Q92736, (Rat) B0LPN4, (Mouse) E9Q401
Entrez Gene ID: (Human) 6262, (Rat) 689560, (Mouse) 20191
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.