Invitrogen
Manufacturer Code:PA5115857
Catalog # PIPA5115857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RYR1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI528790 calcium release channel calcium release channel (ryanodine receptor 1) calcium release channel protein CCO central core disease of muscle CRC CRC1 RYR1 halothane LOW QUALITY PROTEIN: ryanodine receptor 1 MHS MHS1 porcine stress syndrome PPP1R137 protein phosphatase 1 regulatory subunit 137 ryanodine receptor Ryanodine receptor 1 ryanodine receptor 1 (skeletal) ryanodine receptor 1 skeletal muscle ryanodine receptor 1-like skeletal muscle RYDR RYR Ryr1 RYR-1 Ryr1l sarcoplasmic reticulum calcium release channel Skeletal muscle calcium release channel skeletal muscle ryanodine receptor Skeletal muscle-type ryanodine receptor skrr Type 1 ryanodine receptor type 1-like ryanodine receptor; central core disease of muscle; protein phosphatase 1, regulatory subunit 137; Ryanodine receptor 1; ryanodine receptor 1 (skeletal); RYR-1; sarcoplasmic reticulum calcium release channel; Skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; Skeletal muscle-type ryanodine receptor; Type 1 ryanodine receptor; type 1-like ryanodine receptor
Gene Aliases: CCO; MHS; MHS1; PPP1R137; RYDR; RYR; RYR-1; RYR1; SKRR
UniProt ID: (Human) P21817
Entrez Gene ID: (Human) 6261
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.