Invitrogen
Manufacturer Code:PA5111900
Catalog # PIPA5111900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 1371-1386 of human RyR1 (Intracellular N-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI528790 BOS_17377 CCO CRC MHS MHS1 PPP1R137 Ryanodine receptor Ryanodine receptor 1 RYDR RYR RYR1 RYR-1 Ryr1l Skeletal muscle calcium release channel Skeletal muscle ryanodine receptor Skeletal muscle-type ryanodine receptor SKRR Type 1 ryanodine receptor; calcium release channel; central core disease of muscle; protein phosphatase 1, regulatory subunit 137; Ryanodine receptor 1; ryanodine receptor 1 (skeletal); ryanodine receptor 1, skeletal muscle; ryanodine receptor 1-like, skeletal muscle; RYR-1; sarcoplasmic reticulum calcium release channel; Skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; Skeletal muscle-type ryanodine receptor; Type 1 ryanodine receptor; type 1-like ryanodine receptor
Gene Aliases: AI528790; CCO; MHS; MHS1; PPP1R137; RYDR; RYR; RYR-1; RYR1; Ryr1l; SKRR
UniProt ID: (Human) P21817, (Mouse) E9PZQ0
Entrez Gene ID: (Human) 6261, (Mouse) 20190, (Rat) 114207
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.