Abnova
Manufacturer Code:PAB8983
Catalog # 89-134-068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Rpe65. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Rpe65.
For Research Use Only
Protein Aliases: 65kDa; A930029L06Rik; All-trans-retinyl-palmitate hydrolase; LCA2; Lutein isomerase; Meso-zeaxanthin isomerase; modifier of retinal degeneration 1; Mord1; rd12; retinal pigment epithelium, 65 kDa; Retinal pigment epithelium-specific 65 kDa protein; Retinoid isomerohydrolase; Retinol isomerase; RP20
Gene Aliases: 65kDa; A930029L06Rik; LCA2; Mord1; rd12; RP20; Rpe65
UniProt ID: (Mouse) Q91ZQ5
Entrez Gene ID: (Mouse) 19892
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.