Novus Biologicals
Manufacturer Code:NB100655G
Catalog # NB100655G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 500 and 550 of human Ring Finger and CCCH-type finder domains 1 using the numbering given in entry NP_742068.1 (GeneID 149041). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA2025RP5-1198E17.5 RING finger and C3H zinc finger protein 1 ring finger and CCCH-type domains 1 RING finger protein 198 RNF198ring finger and CCCH-type zinc finger domains 1 roquin; probable E3 ubiquitin-protein ligase Roquin; RING finger and C3H zinc finger protein 1; RING finger and CCCH-type zinc finger domain-containing protein 1; RING finger protein 198; Roquin; Roquin-1
Gene Aliases: KIAA2025; RC3H1; RNF198; ROQUIN
UniProt ID: (Human) Q5TC82
Entrez Gene ID: (Human) 149041
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.