Invitrogen
Manufacturer Code:PA5117414
Catalog # PIPA5117414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of mouse ROBO2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2600013A04Rik 9430089E08Rik BB097918 D230004I22Rik KIAA1568 mKIAA1568 ROBO2 roundabout 2 roundabout guidance receptor 2 Roundabout homolog 2 roundabout homolog 2 (Drosophila) roundabout axon guidance receptor homolog 2 roundabout axon guidance receptor homolog 2 (Drosophila) SAX3; roundabout 2; roundabout homolog 2
Gene Aliases: 2600013A04Rik; 9430089E08Rik; BB097918; D230004I22Rik; mKIAA1568
Entrez Gene ID: (Rat) 84409, (Mouse) 268902
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.