Novus Biologicals
Manufacturer Code:NB1001586
Catalog # NB1001586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 1425 and 1450 of human Rap1 Interacting Factor 1 using the numbering given in entry NP_060621.3 (GeneID 55183). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp434D1026 DKFZp781N1478 FLJ10599 FLJ12870 RAP1 interacting factor homolog (yeast) Rap1-interacting factor 1 homolog telomere-associated protein RIF1; RAP1 interacting factor homolog; Rap1-interacting factor 1 homolog; Telomere-associated protein RIF1
Gene Aliases: RIF1
UniProt ID: (Human) Q5UIP0
Entrez Gene ID: (Human) 55183
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.