Abnova
Manufacturer Code:PAB10275
Catalog # 89-114-746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 2406-2419 of mouse Rif1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Rif1.
For Research Use Only
Protein Aliases: 5730435J01Rik; 6530403D07Rik; AU016181; AW549474; D2Ertd145e; mRif1; Rap1-interacting factor 1 homolog; Telomere-associated protein RIF1
Gene Aliases: 5730435J01Rik; 6530403D07Rik; AU016181; AW549474; D2Ertd145e; Rif1
UniProt ID: (Mouse) Q6T8D9
Entrez Gene ID: (Mouse) 51869
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.