Invitrogen
Manufacturer Code:MA515006
Catalog # PIMA515006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | J.644.6 |
Immunogen | Synthetic peptide corresponding to the sequence Gln1681 of human Rictor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AVO3 homolog; AVO3 homolog TORC2-specific protein AVO3 hAVO3 pianissimo rapamycin-insensitive companion of mTOR; FYN binding protein; hAVO3; mAVO3; pianissimo; Protein pianissimo; Rapamycin-insensitive companion of mTOR; RICTOR; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3
Gene Aliases: 4921505C17Rik; 6030405M08Rik; AVO3; AW492497; D530039E11Rik; Fyb; hAVO3; KIAA1999; PIA; RICTOR
UniProt ID: (Human) Q6R327, (Mouse) Q6QI06
Entrez Gene ID: (Human) 253260, (Rat) 310131, (Mouse) 78757
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.