Novus Biologicals
Manufacturer Code:NBP151645
Catalog # NBP151645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7B3 |
Immunogen | Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt# Q6R327] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AVO3 AVO3 homolog hAVO3 KIAA1999DKFZp686B11164 mAVO3 MGC39830 PIA pianissimo rapamycin-insensitive companion of mTOR RPTOR independent companion of MTOR complex 2 TORC2-specific protein AVO3; AVO3 homolog; hAVO3; pianissimo; Rapamycin-insensitive companion of mTOR; RICTOR; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3
Gene Aliases: AVO3; hAVO3; KIAA1999; PIA; RICTOR
UniProt ID: (Human) Q6R327
Entrez Gene ID: (Human) 253260
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.