Invitrogen
Manufacturer Code:PA544605
Catalog # PIPA544605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RPE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: R-phycoerythrin R-PE RPE PE; Ribulose-5-phosphate-3-epimerase; Ribulose-5-phosphate-epimerase; Ribulose-phosphate 3-epimerase
Gene Aliases: 2810429B02Rik; 5730518J08Rik; HUSSY-17; ik:tdsubc_2c8; RPE; RPE2-1; wu:fa07h08; wu:fb93d11; wu:fc20b11; xx:tdsubc_2c8
UniProt ID: (Human) Q96AT9, (Mouse) Q8VEE0
Entrez Gene ID: (Pig) 100522764, (Bovine) 533764, (Dog) 478893, (Horse) 100051899, (Human) 6120, (Rabbit) 100342117, (Guinea pig) 100730209, (Mouse) 66646, (Rat) 501157, (Zebrafish) 334897
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.