Novus Biologicals
Manufacturer Code:NBP2856360.1ML
Catalog # NB033565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human Ribosomal Protein L24 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L24; 60S ribosomal protein L24 60S ribosomal protein L30 L24 ribosomal protein L30 RPL24; 60S ribosomal protein L30; epididymis secretory protein Li 310; Large ribosomal subunit protein eL24; ribosomal protein L30
Gene Aliases: HEL-S-310; L24; RPL24
UniProt ID: (Human) P83731
Entrez Gene ID: (Human) 6152
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.