Novus Biologicals
Manufacturer Code:NBP273896AF647
Catalog # N273896A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B7 |
Immunogen | Full length recombinant protein of human Ribonuclease Inhibitor (NP_002930) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Placental ribonuclease inhibitor; Placental ribonuclease inhibitor Placental RNase inhibitor PRI RAIMGC4569 ribonuclease inhibitor ribonuclease/angiogenin inhibitor ribonuclease/angiogenin inhibitor 1MGC18200 RNHMGC54054; Placental RNase inhibitor; RAI; Ribonuclease inhibitor; Ribonuclease/angiogenin inhibitor 1; testicular tissue protein Li 164
Gene Aliases: PRI; RAI; RNH; RNH1
UniProt ID: (Human) P13489
Entrez Gene ID: (Human) 6050
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.