Novus Biologicals
Manufacturer Code:NBP281059AF750
Catalog # NB015100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | Rho 1D4 |
Immunogen | Bleached bovine rod outer segment (ROS). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CSNBAD1 MGC138309 OPN2MGC138311 opsin 2 rod pigment opsin-2 retinitis pigmentosa 4 autosomal dominant rhodopsin RP4; opsin 2, rod pigment; Opsin-2; Rhodopsin
Gene Aliases: CSNBAD1; OPN2; RHO; RP4
UniProt ID: (Human) P08100
Entrez Gene ID: (Human) 6010
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.