Invitrogen
Manufacturer Code:MA511738
Catalog # PIMA511738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | RET-P1 |
Immunogen | Membrane preparation from adult rat retina |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: L opsin; Long Wavelength Sensitive opsin; LWS opsin; opsin 2; opsin 2 rod pigment opsin-2 retinitis pigmentosa 4 autosomal domit rhodopsin (opsin 2 rod pigment) (retinitis pigmentosa 4 autosomal domit); opsin 2, rod pigment; Opsin-2; Red Opsin; Rhodopsin; Rhodopsin (retinitis pigmentosa 4, autosomal dominant); Rod Opsin
Gene Aliases: CSNBAD1; Noerg1; OPN2; Ops; RHO; RP4
UniProt ID: (Human) P08100, (Rat) P51489, (Mouse) P15409
Entrez Gene ID: (Human) 6010, (Rat) 24717, (Mouse) 212541
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.