Invitrogen
Manufacturer Code:PA572238
Catalog # PIPA572238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 62-93 amino acids from the human region of DANRE rho |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: retinal rod opsin pigment rh1.1; Rhodopsin; Rhodopsin rho zfo2; rhodopsin-like-1.1 rod opsin; rod opsin; seven transmembrane light receptor
Gene Aliases: Rh; RH1; rh1.1; rho; wu:fi06d11; zfo2; zfrho
UniProt ID: (Zebrafish) P35359
Entrez Gene ID: (Zebrafish) 30295
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.