Invitrogen
Manufacturer Code:PA551809
Catalog # PIPA551809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RhoJ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ras homolog gene family, member J; Ras-like protein family member 7B; RAS-like, family 7, member B; Rho-related GTP-binding protein RhoJ; Rho-related GTP-binding protein RhoJ Ras-like protein family member 7B Tc10-like GTP-binding protein RHOJ ARHJ RASL7B RHOI TCL; Tc10-like GTP-binding protein; TC10-like Rho GTPase
Gene Aliases: ARHJ; RASL7B; RHOI; RHOJ; TC10B; TCL
UniProt ID: (Human) Q9H4E5
Entrez Gene ID: (Human) 57381
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.