Novus Biologicals
Manufacturer Code:NBP237472
Catalog # NBP237472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G3 |
Immunogen | Purified recombinant fragment of human ARHGDIA (AA: FULL(1-204)) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory sperm binding protein Li 47e; GDIA1 MGC117248 Rho GDI 1 Rho GDP dissociation inhibitor (GDI) alpha rho GDP-dissociation inhibitor 1 RHOGDI Rho-GDI alpha RHOGDI-1; GDP-dissociation inhibitor, aplysia RAS-related 1; Rho GDI 1; Rho GDP dissociation inhibitor (GDI) alpha; Rho GDP-dissociation inhibitor 1; Rho-GDI alpha
Gene Aliases: ARHGDIA; GDIA1; HEL-S-47e; NPHS8; RHOGDI; RHOGDI-1
UniProt ID: (Human) P52565
Entrez Gene ID: (Human) 396
Molecular Function: G-protein modulator enzyme modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.