Invitrogen
Manufacturer Code:PA5104436
Catalog # PIPA5104436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RHG4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A130009C12Rik A530015A20Rik ARHGAP4 C1 GTPase-activating protein p115 KIAA0131 mKIAA0131 p115 RG RhoGAP4 RGC1 Rho GTPase activating protein 4 rho GTPase-activating protein 4 Rho-GAP hematopoietic protein C1 RHOGAP4 rho-type GTPase-activating protein 4 SrGAP4; GTPase-activating protein p115; p115; Rho GTPase-activating protein 4; Rho-GAP hematopoietic protein C1; Rho-type GTPase-activating protein 4
Gene Aliases: A130009C12Rik; A530015A20Rik; ARHGAP4; C1; KIAA0131; mKIAA0131; p115; RGC1; RHOGAP4; SrGAP4
UniProt ID: (Human) Q86UY3
Entrez Gene ID: (Human) 393, (Mouse) 171207, (Rat) 246249
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.