Novus Biologicals
Manufacturer Code:NBP220156
Catalog # NBP220156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human RhoC. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARH9ARHCRho cDNA clone 9 H9 MGC1448 MGC61427 oncogene RHO H9 ras homolog gene family member C RAS-related homolog 9 RhoC rhoC GTPase RHOH9 rho-related GTP-binding protein RhoC small GTP binding protein RhoC; h9; oncogene RHO H9; ras homolog gene family, member C; RAS-related homolog 9; Rho cDNA clone 9; Rho-related GTP-binding protein RhoC; rhoC GTPase; small GTP binding protein RhoC
Gene Aliases: ARH9; ARHC; H9; RHOC; RHOH9
UniProt ID: (Human) P08134
Entrez Gene ID: (Human) 389
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.