Invitrogen
Manufacturer Code:PA588525
Catalog # PIPA588525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-196 of human RHOB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aplysia ras-related homolog B; ras homolog B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho-related GTP-binding protein RhoB; rhoB
Gene Aliases: AA017882; Arh6; Arhb; Rhob
UniProt ID: (Rat) P62747, (Mouse) P62746
Entrez Gene ID: (Rat) 64373, (Mouse) 11852
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.