Invitrogen
Manufacturer Code:PA5115844
Catalog # PIPA5115844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RHOB corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA017882 Aplysia RAS-related homolog 6 aplysia ras-related homolog B Arh6 Arhb h6 MST081 MSTP081 oncogene RHO H6 ras homolog B ras homolog family member B ras homolog gene family member AB ras homolog gene family member B Rho rho cDNA clone 6 Rhob RHOH6 Rho-related GTP-binding protein RhoB; Aplysia RAS-related homolog 6; aplysia ras-related homolog B; h6; oncogene RHO H6; ras homolog B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho cDNA clone 6; Rho-related GTP-binding protein RhoB; rhoB
Gene Aliases: AA017882; ARH6; ARHB; MST081; MSTP081; RHOB; RHOH6
UniProt ID: (Human) P62745, (Rat) P62747, (Mouse) P62746
Entrez Gene ID: (Human) 388, (Rat) 64373, (Mouse) 11852
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.