Novus Biologicals
Manufacturer Code:NBP1034840.1MG
Catalog # NB018269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide as a part of human rat mouse Rho-related GTP-binding protein RhoB (ARH6 ARHB H6) conjugated to an immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aplysia RAS-related homolog 6; ARH6MSTP081 ARHBAplysia RAS-related homolog 6 h6 MST081 oncogene RHO H6 ras homolog gene family member B Rho cDNA clone 6 RHOH6RhoB rho-related GTP-binding protein RhoB; h6; oncogene RHO H6; ras homolog gene family, member B; Rho cDNA clone 6; Rho-related GTP-binding protein RhoB
Gene Aliases: ARH6; ARHB; MST081; MSTP081; RHOB; RHOH6
UniProt ID: (Human) P62745
Entrez Gene ID: (Human) 388
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.