Invitrogen
Manufacturer Code:MA1123
Catalog # PIMA1123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B3-4A10 |
Immunogen | Full-length human recombit RhoA protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aplysia ras-related homolog 12; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; h12; h9; oncogene RHO H12; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member C; RAS-related homolog 9; Rho cDNA clone 12; Rho cDNA clone 9; Rho family GTPase; Rho-related GTP-binding protein RhoC; Rhoa RHOC ARH12 ARHA RHO12; rhoC GTPase; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; Transforming protein RhoA
Gene Aliases: AI324259; ARH12; ARH9; ARHA; Arha1; Arha2; ARHC; H9; RHO12; RHOA; RHOC; RHOH12; RHOH9
UniProt ID: (Human) P61586, (Human) P08134, (Rat) P61589, (Mouse) Q9QUI0, (Mouse) Q62159
Entrez Gene ID: (Human) 387, (Human) 389, (Rat) 117273, (Mouse) 11848, (Mouse) 11853
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.