Invitrogen
Manufacturer Code:MA1011
Catalog # PIMA1011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A11-4G10 |
Immunogen | Full-length recombit human Rho A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; aplysia ras-related homolog B; h12; h6; h9; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog B; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member AB; ras homolog gene family, member B; ras homolog gene family, member C; RAS-related homolog 9; Rho cDNA clone 12; Rho cDNA clone 6; Rho cDNA clone 9; Rho family GTPase; Rho-related GTP-binding protein RhoB; Rho-related GTP-binding protein RhoC; Rhoa Rhob RHOC ARH12 ARHA RHO12 Rho cDNA clone 12; rhoB; rhoC GTPase; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; Transforming protein RhoA
Gene Aliases: AA017882; AI324259; ARH12; ARH6; ARH9; ARHA; Arha1; Arha2; ARHB; ARHC; H9; MST081; MSTP081; RHO12; RHOA; RHOB; RHOC; RHOH12; RHOH6; RHOH9
UniProt ID: (Human) P61586, (Human) P62745, (Human) P08134, (Rat) P61589, (Mouse) Q9QUI0, (Rat) P62747, (Mouse) P62746, (Mouse) Q62159
Entrez Gene ID: (Human) 387, (Human) 388, (Human) 389, (Rat) 117273, (Mouse) 11848, (Rat) 64373, (Mouse) 11852, (Mouse) 11853, (Rat) 295342
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.