Novus Biologicals
Manufacturer Code:NBP290107100UL
Catalog # NB021677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 103 |
Immunogen | Recombinant Human RhoA protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Aplysia ras-related homolog 12; h12; h12 oncogene RHO H12 ras homolog gene family member A Rho cDNA clone 12 Rho12 RhoA RHOH12ARH12ARHAAplysia ras-related homolog 12 small GTP binding protein RhoA transforming protein RhoA; oncogene RHO H12; Rho cDNA clone 12; small GTP binding protein RhoA; Transforming protein RhoA
Gene Aliases: ARH12; ARHA; RHO12; RHOA; RHOH12
UniProt ID: (Human) P61586
Entrez Gene ID: (Human) 387
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.