Invitrogen
Manufacturer Code:PA577864
Catalog # PIPA577864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human RhoA. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aplysia ras-related homolog 12; aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; ARH12 ARHA RHO12 RHOH12 RHOA oncogene RHO H12 Aplysia ras-related homolog 12 rho cDNA clone 12 h12 small GTP binding protein RhoA transforming protein RhoA ras homolog gene family member A; h12; oncogene RHO H12; Ras family member A; ras homolog A1; ras homolog A2; ras homolog gene family, member A1; ras homolog gene family, member A2; Rho cDNA clone 12; Rho family GTPase; small GTP binding protein RhoA; Transforming protein RhoA
Gene Aliases: ARH12; ARHA; Arha1; Arha2; RHO12; RHOA; RHOH12
UniProt ID: (Human) P61586, (Mouse) Q9QUI0
Entrez Gene ID: (Human) 387, (Mouse) 11848
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.