Novus Biologicals
Manufacturer Code:NBP267325
Catalog # NBP267325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SR38-00 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Aplysia ras-related homolog 12; Aplysia ras-related homolog 12 Aplysia RAS-related homolog 6 ARH12 ARH6 ARH9 ARHA ARHB ARHC EDFAOB epididymis secretory sperm binding protein h6 H9 MST081 MSTP081 oncogene RHO H12 oncogene RHO H6 oncogene RHO H9 ras homolog gene family member B ras homolog gene family member C RAS-related homolog 9 Rho cDNA clone 6 RHO12 rhoC GTPase RHOH12 RHOH6 RHOH9 rho-related GTP-binding protein RhoB rho-related GTP-binding protein RhoC small GTP binding protein RhoA small GTP binding protein RhoC transforming protein RhoA; h12; oncogene RHO H12; Rho cDNA clone 12; small GTP binding protein RhoA; Transforming protein RhoA
Gene Aliases: ARH12; ARHA; RHO12; RHOA; RHOH12
UniProt ID: (Human) P61586
Entrez Gene ID: (Human) 387
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.