Novus Biologicals
Manufacturer Code:NBP158861
Catalog # NBP158861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RHEB(Ras homolog enriched in brain) The peptide sequence was selected from the middle region of RHEB. Peptide sequence VGKVQIPIMLVGNKKDLHMERVISYEEGKALAESWNAAFLESSAKENQTA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein Rheb; GTP-binding protein Rheb MGC111559 Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2; Ras homolog enriched in brain; Ras homolog enriched in brain 2
Gene Aliases: RHEB; RHEB2
UniProt ID: (Human) Q15382
Entrez Gene ID: (Human) 6009
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.