Novus Biologicals
Manufacturer Code:NBP237357
Catalog # NBP237357
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5E11A6 |
Immunogen | Purified recombinant fragment of human ZFP42 (AA: 249-310) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ20086 KIAA1584 MGC21637 MGC61687 SUHW4 Suppressor of hairy wing homolog 4 zinc finger protein 280D Zinc finger protein 634 ZNF634suppressor of hairy wing homolog 4 (Drosophila); Reduced expression protein 1; REX-1; REX1 transcription factor; Zfp-42; Zinc finger protein 42 homolog; Zinc finger protein 754
Gene Aliases: REX-1; REX1; zfp-42; ZFP42; ZNF754
UniProt ID: (Human) Q96MM3
Entrez Gene ID: (Human) 132625
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.