R&D Systems
Manufacturer Code:PPA8740A00
Catalog # PPA8740A00
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A8740A |
Immunogen | Recombinant human Rev-Erb alpha/NR1D1 aa 2-130 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | This antibody is stable for greater than six months when held at -20degrees C in a manual defrost freezer or at -70degrees C. Upon thawing the antibody can be stored at 2degrees - 8degrees C for at least 1 month without detectable loss of activity. A |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EAR-1; EAR-1 EAR1nuclear receptor subfamily 1 group D member 1 HREV NR1D1 nuclear receptor subfamily 1 group D member 1 Reverb A alpha Rev-erbA-alpha Rev-ErbAalpha THRA1 THRAL THRALrev-erbA-alpha V-erbA-related protein 1; nuclear receptor Rev-ErbA-alpha; Nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; Rev-erbA-alpha; Rev-ErbAalpha; V-erbA-related protein 1
Gene Aliases: ear-1; EAR1; HREV; NR1D1; THRA1; THRAL
UniProt ID: (Human) P20393
Entrez Gene ID: (Human) 9572
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.