Novus Biologicals
Manufacturer Code:NBP15767720UL
Catalog # NBP15767720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RBP1(retinol binding protein 1 cellular) The peptide sequence was selected from the middle region of RBP1. Peptide sequence IIRTLSTFRNYIMDFQVGKEFEEDLTGIDDRKCMTTVSWDGDKLQCVQKG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C Cellular retinol-binding protein CRABP-I CRBP1CRBP-I CRBPCellular retinol-binding protein I CRBPI RBPC retinol binding protein 1 cellular retinol-binding protein 1 retinol-binding protein 1 cellular; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRBP; CRBP-I; Retinol-binding protein 1; retinol-binding protein 1, cellular
Gene Aliases: CRABP-I; CRBP; CRBP1; CRBPI; RBP1; RBPC
UniProt ID: (Human) P09455
Entrez Gene ID: (Human) 5947
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.