Novus Biologicals
Manufacturer Code:NB200202
Catalog # NB200202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Reacts with residues 94-102 [AGMDWTGAR] of the human Resistin protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adipose tissue-specific secretory factor; Adipose tissue-specific secretory factor ADSFMGC126609 C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1 C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein Cysteine-rich secreted protein A12-alpha-like 2 Cysteine-rich secreted protein FIZZ3 FIZZ3MGC126603 found in inflammatory zone 3 HXCP1 resistin RETN1 RSTNXCP1; ADSF; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 1; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; Cysteine-rich secreted protein A12-alpha-like 2; Cysteine-rich secreted protein FIZZ3; found in inflammatory zone 3; Resistin; resistin delta2
Gene Aliases: ADSF; FIZZ3; HXCP1; RETN; RETN1; RSTN; UNQ407/PRO1199; XCP1
UniProt ID: (Human) Q9HD89
Entrez Gene ID: (Human) 56729
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.