Novus Biologicals
Manufacturer Code:NBP273851AF700
Catalog # N273851A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F10 |
Immunogen | Recombinant protein fragment corresponding to amino acids 131-406 of human Renin (NP_000528) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: angiotensin-forming enzyme; angiotensin-forming enzyme Angiotensinogenase EC 3.4.23 EC 3.4.23.15 FLJ10761 HNFJ2 Prorenin renin renin precursor renal; Angiotensinogenase; Renin; renin precursor, renal
Gene Aliases: HNFJ2; REN
UniProt ID: (Human) P00797
Entrez Gene ID: (Human) 5972
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.