Novus Biologicals
Manufacturer Code:NBP289723100UL
Catalog # NB020927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 9 |
Immunogen | Recombinant Human Renin protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: angiotensin-forming enzyme; angiotensin-forming enzyme Angiotensinogenase EC 3.4.23 EC 3.4.23.15 FLJ10761 HNFJ2 renin renin precursor renal; Angiotensinogenase; Renin; renin precursor, renal
Gene Aliases: HNFJ2; REN
UniProt ID: (Human) P00797
Entrez Gene ID: (Human) 5972
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.