Invitrogen
Manufacturer Code:PA547577
Catalog # PIPA547577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Renin Leu24-Arg406 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: angiotensin-forming enzyme; Angiotensinogenase; HNFJ2 angiotensin-forming enzyme angiotensinogenase renin precursor renal; Renin; renin precursor, renal
Gene Aliases: HNFJ2; REN
UniProt ID: (Human) P00797
Entrez Gene ID: (Human) 5972
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.