Invitrogen
Manufacturer Code:PA521690
Catalog # PIPA521690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 70 and 332 of Renin (Uniprot IDNo.P00797) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: angiotensin-forming enzyme; Angiotensinogenase; HNFJ2 angiotensin-forming enzyme angiotensinogenase renin precursor renal; Renin; renin 1 structural; renin precursor, renal
Gene Aliases: HNFJ2; RATRENAA; REN; Ren1; RENAA
UniProt ID: (Human) P00797, (Rat) P08424
Entrez Gene ID: (Human) 5972, (Rat) 24715
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.