Invitrogen
Manufacturer Code:PA518736
Catalog # PIPA518736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QINLRDDKWEVSKQ) corresponding to the internal amino acids of C10orf59 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-NAD(P)H oxidase/anomerase; MAO-C; mMAO-C; Monoamine oxidase-C; Renalase; renalase C10orf59 chromosome 10 open reading frame 59 FLJ11218; rnls
Gene Aliases: 6530404N21Rik; AI452315; AW060440; C10orf59; RGD1309804; Rnls
UniProt ID: (Rat) Q5U2W9
Entrez Gene ID: (Rat) 361751, (Mouse) 67795
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.